Merck
  • Small-molecule inhibition of aging-associated chromosomal instability delays cellular senescence.
Small-molecule inhibition of aging-associated chromosomal instability delays cellular senescence.
EMBO reports (2020-03-07)
Monika Barroso-Vilares, Joana C Macedo, Marta Reis, Jessica D Warren, Duane Compton, Elsa Logarinho
ABSTRACT
Chromosomal instability (CIN) refers to the rate at which cells are unable to properly segregate whole chromosomes, leading to aneuploidy. Besides its prevalence in cancer cells and postulated implications in promoting tumorigenesis, studies in aneuploidy-prone mouse models uncovered an unanticipated link between CIN and aging. Using young to old-aged human dermal fibroblasts, we observed a dysfunction of the mitotic machinery arising with age that mildly perturbs chromosome segregation fidelity and contributes to the generation of fully senescent cells. Here, we investigated mitotic mechanisms that contribute to age-associated CIN. We found that elderly cells have an increased number of stable kinetochore-microtubule (k-MT) attachments and decreased efficiency in the correction of improper k-MT interactions. Chromosome mis-segregation rates in old-aged cells decreased upon both genetic and small-molecule enhancement of MT-depolymerizing kinesin-13 activity. Notably, restored chromosome segregation accuracy inhibited the phenotypes of cellular senescence. Therefore, we provide mechanistic insight into age-associated CIN and disclose a strategy for the use of a small-molecule to inhibit age-associated CIN and to delay the cellular hallmarks of aging.
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein di(β-D-galactopyranoside)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).