Merck
  • Contrast and efficiency enhancement in organic light-emitting devices utilizing high absorption and high charge mobility organic layers.
Contrast and efficiency enhancement in organic light-emitting devices utilizing high absorption and high charge mobility organic layers.
Optics express (2006-08-21)
W F Xie, Y Zhao, C N Li, S Y Liu
ABSTRACT
Organic light-emitting devices utilizing copper phthalocyanine (CuPc) and C(60) as the hole- and electron-injection layers were reported. Compared with a conventional device without CuPc and C(60) layers, the improvement of the contrast is more than 100% under 140-lx ambient lighting at a brightness of 300 cd/m(2). A maximum current efficiency of 3.93 cd/A, which is higher than 3.62 cd/A for a conventional device, was obtained at 9 V. The device has a maximum luminance of 17170 cd/m(2) at 15 V. The high contrast and high efficiency of the device are attributed to the high absorption and high charge mobility of CuPc and C(60) films.
Sigma-Aldrich
Copper(II) phthalocyanine, sublimed grade, Dye content 99 %
Sigma-Aldrich
Copper(II) phthalocyanine, Dye content >99 %
Sigma-Aldrich
Copper(II) phthalocyanine, β-form, Dye content 90 %
Sigma-Aldrich
Copper(II) phthalocyanine, triple-sublimed grade, >99.95% trace metals basis
Sigma-Aldrich
Fullerene-C60, 99.5%
Sigma-Aldrich
Fullerene-C60, 98%
Sigma-Aldrich
Fullerene-C60, sublimed, 99.9%