Merck
  • Core-shell pea protein-carboxymethylated corn fiber gum composite nanoparticles as delivery vehicles for curcumin.
Core-shell pea protein-carboxymethylated corn fiber gum composite nanoparticles as delivery vehicles for curcumin.
Carbohydrate polymers (2020-06-02)
Yue Wei, Zhixiang Cai, Min Wu, Yalong Guo, Pengguang Wang, Ruiqi Li, Aiqin Ma, Hongbin Zhang
ABSTRACT
Nanocomposites from plant-derived polysaccharides and proteins are green and sustainable materials that can be applied in various food and biomedical fields. In this work, we developed a novel core-shell nanocomposite from carboxymethylated corn fiber gum (CMCFG) and high nutritious pea protein (PP) for curcumin (Cur) delivery. In the preparation, PP-Cur complexes (PP-Cur) were formed at pH 7.0 and then coated by CMCFG via hydrophobic interactions to form PP-Cur-CMCFG complexes. Furthermore, the interactions between CMCFG and PP-Cur in the complexes were enhanced at pH 3.5 by electrostatic interactions. The resulting nanocomposite exhibited an excellent encapsulation performance for Cur with high Cur loading efficiency, good water dispersibility, and high chemical and thermal stability. Besides, the Cur-loaded core-shell nanocomposite also showed higher antioxidant and radical scavenging activities than Cur. Our results provide support for the use of plant-derived delivery systems as a strategy for the delivery of chemically unstable hydrophobic Cur.
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Pancreatin from porcine pancreas, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein