Merck
  • MAML1 promotes ESCC aggressiveness through upregulation of EMT marker TWIST1.
MAML1 promotes ESCC aggressiveness through upregulation of EMT marker TWIST1.
Molecular biology reports (2020-03-18)
Mohammad Mahdi Forghanifard, Shirin Azaraz, Sima Ardalan Khales, Dorsa Morshedi Rad, Mohammad Reza Abbaszadegan
ABSTRACT
Mastermind-like 1 (MAML1) is the main transcriptional co-activator of Notch signaling pathway. It plays essential roles in several pathways including MEF2C, p53, Nf-кB and Wnt/β-catenin. TWIST1 is known as a regulator of epithelial mesenchymal transition (EMT), which is considered as a primary step in promotion of tumor cell metastasis. Since concomitant expression of these genes was observed in tumors, our aim in this study was to elucidate the linkage between MAML1 and TWIST1 co-overexpression in esophageal squamous cell carcinoma (ESCC). While MAML1 silencing significantly down-regulated TWIST1, its ectopic expression up-regulated TWIST1 expression in both mRNA and protein levels in KYSE-30 cells. Expression of mesenchymal markers was increased significantly after MAML1 and TWIST1 ectopic expression, while epithelial markers expression was significantly decreased after silencing of both genes. Concomitant protein expression of MAML1 and TWIST1 was significantly observed in ESCC patients. Enforced expression of TWIST1 had no impact on MAML1 gene expression in KYSE-30 cells. The results clearly suggest transcriptional regulation of TWIST1 by MAML1 transcription factor in ESCC cells KYSE-30. Since TWIST1 is known as an EMT inducing marker, our results may revealed the mastermind behind TWIST1 function and introduced MAML1 as an upstream master regulator of TWIST1 and EMT in KYSE-30 cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAML1