Merck
  • Fabrication of new generation of co-delivery systems based on graphene-g-cyclodextrin/chitosan nanofiber.
Fabrication of new generation of co-delivery systems based on graphene-g-cyclodextrin/chitosan nanofiber.
International journal of biological macromolecules (2019-11-22)
Shabnam Sattari, Abbas Dadkhah Tehrani, Mohsen Adeli, Khadijeh Soleimani, Marzieh Rashidipour
ABSTRACT
Although many techniques have been devoted to promote therapeutic purposes of drug carrier systems, however, there are still many challenges in this area. Here, we designed co-loaded delivery systems, composed of curcumin loaded cyclodextrin-graphene oxide core (Cur@CD-GO) and gallic acid loaded chitosan shell nanofibers (Cur-Ga NF), which can promote the therapeutic efficiency of drugs. The synthesized nanofibres were fabricated by electrospinning technique with the coaxial system. Results showed that co-loaded delivery systems (Cur-Ga NF) provide better performance over single drug-loaded NFs (Cur@CD-GO). It was demonstrated that the nanofibers were successfully prepared, and the drugs in the core and sell of nanofibers were released in a controlled and sustained manner. The produced Cur-Ga NF, providing improved anti-cancer activity, antimicrobial activity, antioxidant activity and anti-inflammatory outcome as compared to single drug-loaded NFs. Our investigations showed that such co-delivery fiber systems could be employed as a promising nanocarrier for therapeutic applications.
Sigma-Aldrich
2-Ethyl-2-oxazoline, ≥99%