Merck
  • Antimicrobial and antiproliferative activity studies of some new quinoline-3-carbaldehyde hydrazone derivatives.
Antimicrobial and antiproliferative activity studies of some new quinoline-3-carbaldehyde hydrazone derivatives.
Bioorganic chemistry (2020-07-01)
Mustafa Orhan Puskullu, Ismail Celik, Meryem Erol, Hanifa Fatullayev, Ebru Uzunhisarcikli, Gulcan Kuyucuklu
ABSTRACT
In this study, a total of 22 piece quinoline-3-carbaldehyde hydrazone derivative compounds were designed and synthesized, 2 of which were not original, their antimicrobial activities were determined with microdilution method and their in vitro cytotoxic effect was investigated in MCF-7 and A549 cells by MTT assay. When the activity results are examined, although the antimicrobial effects of quinoline derivatives, in general, are weaker than standard drugs; 3q5 and 3q6 against MRSA showed promising activity with MIC:16 µg/ml compared to reference drugs. Compounds generally showed weaker cytotoxic activity on the A549 and MCF-7 cell line. 3q12, 3q17 and 3q22 at 100 µM reduced cell viability to 59.28%, 76.24% and 72.92% on A549 cells, respectively. Compound 3q6, one of the most effective compounds against MRSA, formed a 2.10 Å long hydrogen bond between the quinoline nitrogen and ARG132 in the DNA topoisomerase IV active site (PDB: 3FV5). Theoretical ADME profiles of all compounds comply with Lipinski and other limiting rules. In addition, MEP analysis of 3q6, geometric optimization and molecular reactivity analysis were calculated with the 6-311G (d,p) base set DFT/B3LYP theory, and ΔE = ELUMO-EHOMO, which is a measure of the stable structure of the molecule, was calculated as 0.13377 for 3q6 and had the most stable electronic structure among all compounds.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Nutrient Mixture F-12 Ham, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture