Merck
  • Hierarchical Representations of Aggression in a Hypothalamic-Midbrain Circuit.
Hierarchical Representations of Aggression in a Hypothalamic-Midbrain Circuit.
Neuron (2020-03-14)
Annegret L Falkner, Dongyu Wei, Anjeli Song, Li W Watsek, Irene Chen, Patricia Chen, James E Feng, Dayu Lin
ABSTRACT
Although the ventromedial hypothalamus ventrolateral area (VMHvl) is now well established as a critical locus for the generation of conspecific aggression, its role is complex, with neurons responding during multiple phases of social interactions with both males and females. It has been previously unclear how the brain uses this complex multidimensional signal and coordinates a discrete action: the attack. Here, we find a hypothalamic-midbrain circuit that represents hierarchically organized social signals during aggression. Optogenetic-assisted circuit mapping reveals a preferential projection from VMHvlvGlut2 to lPAGvGlut2 cells, and inactivation of downstream lPAGvGlut2 populations results in aggression-specific deficits. lPAG neurons are selective for attack action and exhibit short-latency, time-locked spiking relative to the activity of jaw muscles during biting. Last, we find that this projection conveys male-biased signals from the VMHvl to downstream lPAGvGlut2 neurons that are sensitive to features of ongoing activity, suggesting that action selectivity is generated by a combination of pre- and postsynaptic mechanisms.
Sigma-Aldrich
Clozapine N-oxide
Sigma-Aldrich
Lidocaine N-ethyl chloride
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor