- A 10-year survey of antifungal susceptibility of candidemia isolates from intensive care unit patients in Greece.
A 10-year survey of antifungal susceptibility of candidemia isolates from intensive care unit patients in Greece.
Antimicrobial agents and chemotherapy (2008-12-31)
George Dimopoulos, Aristea Velegraki, Matthew E Falagas
PMID19114672
ABSTRACT
This study retrospectively reviews the susceptibility of 135 baseline ICU candidemia isolates (from 1997 to 2007) to nine antifungals as determined by the AFST-EUCAST microdilution method and identifies the most frequent causative agents of confirmed point-source candidemia outbreaks in local intensive care units. A minority of common and rare Candida species displayed decreased susceptibility to all antifungals.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture