Merck
  • Quorum sensing in Vibrio harveyi: probing the specificity of the LuxP binding site.
Quorum sensing in Vibrio harveyi: probing the specificity of the LuxP binding site.
Bioorganic & medicinal chemistry letters (2005-04-20)
Colin A Lowery, Kathleen M McKenzie, Longwu Qi, Michael M Meijler, Kim D Janda
ABSTRACT
Quorum sensing activity was investigated in the bacterium Vibrio harveyi using a series of both natural and nonnatural analogs of DPD, the penultimate precursor to autoinducer AI-2. The progression of molecules that were both synthesized and investigated includes enantiomeric variants, carbon-chain extension, and hydroxyl-functional group addition/deletions of DPD. The compilation of these studies reveals a binding cleft that can accommodate a number of different structural variants of DPD, albeit with invariably lower activities.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline