Merck
  • Chemical constituents of the fruits of Morinda citrifolia (Noni) and their antioxidant activity.
Chemical constituents of the fruits of Morinda citrifolia (Noni) and their antioxidant activity.
Journal of natural products (2005-04-23)
Bao-Ning Su, Alison D Pawlus, Hyun-Ah Jung, William J Keller, Jerry L McLaughlin, A Douglas Kinghorn
ABSTRACT
Purification of a n-BuOH-soluble partition of the MeOH extract of Morinda citrifolia (Noni) fruits led to the isolation of two new iridoid glucosides, 6alpha-hydroxyadoxoside (1) and 6beta,7beta-epoxy-8-epi-splendoside (2), as well as 17 known compounds, americanin A (3), narcissoside (4), asperuloside, asperulosidic acid, borreriagenin, citrifolinin B epimer a, citrifolinin B epimer b, cytidine, deacetylasperuloside, dehydromethoxygaertneroside, epi-dihydrocornin, d-glucose, d-mannitol, methyl alpha-d-fructofuranoside, methyl beta-d-fructofuranoside, nicotifloroside, and beta-sitosterol 3-O-beta-d-glucopyranoside. The structures of the new compounds were determined by spectroscopic data interpretation. Compound 4, borreriagenin, cytidine, deacetylasperuloside, dehydromethoxygaertneroside, epi-dihydrocornin, methyl alpha-d-fructofuranoside, and methyl beta-d-fructofuranoside were isolated for the first time from M. citrifolia. The antioxidant activity was evaluated for all isolates in terms of both DPPH and ONOO(-) bioassays. The neolignan, americanin A (3), was found to be a potent antioxidant in these assays.
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
D-Penicillamine, 98-101%
Sigma-Aldrich
Asperuloside