Merck
  • SAR studies of 2-methoxyestradiol and development of its analogs as probes of anti-tumor mechanisms.
SAR studies of 2-methoxyestradiol and development of its analogs as probes of anti-tumor mechanisms.
Bioorganic & medicinal chemistry letters (2006-05-03)
Abby Ho, Yang-eon Kim, Hyosung Lee, Kedra Cyrus, Sun-Hee Baek, Kyung-Bo Kim
ABSTRACT
The major estrogen metabolite 2-methoxyestradiol (2ME) has been shown to target tumor cells without severe side effects and is currently being evaluated in clinical trials for several types of cancer. Despite its promise for use in clinical setting, the mechanism(s) by which 2ME exerts its anti-tumor activity is not clearly defined at this time. Employing organic chemistry tools, we synthesized 2ME analogs with which 2ME affinity column was prepared, enabling us to detect a protein that selectively interacts with 2ME. This 2ME analog will be useful as a probe to identify the biological target(s) of 2ME and study their functions in tumor cells.
Sigma-Aldrich
2-Methoxyestradiol, powder