Merck
  • Free radical scavengers from the medicinal mushroom Inonotus xeranticus and their proposed biogenesis.
Free radical scavengers from the medicinal mushroom Inonotus xeranticus and their proposed biogenesis.
Bioorganic & medicinal chemistry letters (2006-08-16)
In-Kyoung Lee, Jin-Young Jung, Soon-Ja Seok, Wan-Gyu Kim, Bong-Sik Yun
ABSTRACT
New free radical scavengers, inoscavin D (1) and methylinoscavin D (2), were isolated from the methanolic extract of the fruiting bodies of Inonotus xeranticus (Hymenochaetaceae), along with the known compounds phelligridin D (3), 3,4-dihydroxybenzaldehyde (4), and 3,4-dihydroxybenzoic acid (5). Their structures were established by various spectroscopic analyses. Compounds 1 and 3 were proposed to be biosynthesized from the oxidative coupling of the precursor hispidin with 3,4-dihydroxybenzaldehyde and 3,4-dihydroxybenzoic acid, respectively. These compounds exhibited significant scavenging activity against the ABTS radical cation, and compounds 2 and 4 displayed moderate superoxide radical scavenging activity.
Sigma-Aldrich
3,4-Dihydroxybenzaldehyde, 97%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)