Merck
  • Molecular mechanism of flucytosine resistance in Candida lusitaniae: contribution of the FCY2, FCY1, and FUR1 genes to 5-fluorouracil and fluconazole cross-resistance.
Molecular mechanism of flucytosine resistance in Candida lusitaniae: contribution of the FCY2, FCY1, and FUR1 genes to 5-fluorouracil and fluconazole cross-resistance.
Antimicrobial agents and chemotherapy (2006-10-25)
Nicolas Papon, Thierry Noël, Martine Florent, Stéphanie Gibot-Leclerc, Dorothée Jean, Christiane Chastin, Jean Villard, Florence Chapeland-Leclerc
ABSTRACT
Inactivation of the FCY2 (cytosine permease), FCY1 (cytosine deaminase), and FUR1 (uracil phosphoribosyltransferase) genes in Candida lusitaniae produced two patterns of resistance to flucytosine. Mutant fur1 demonstrated resistance to 5-fluorouracil, whereas mutants fcy1 and fcy2 demonstrated fluconazole resistance in the presence of subinhibitory flucytosine concentrations.
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications