Merck
  • Substrate competition studies using whole-cell accumulation assays with the major tripartite multidrug efflux pumps of Escherichia coli.
Substrate competition studies using whole-cell accumulation assays with the major tripartite multidrug efflux pumps of Escherichia coli.
Antimicrobial agents and chemotherapy (2007-01-11)
Christopher A Elkins, Lisa B Mullis
ABSTRACT
AcrAB-TolC is the major, constitutively expressed tripartite multidrug efflux system in Escherichia coli that recognizes various structurally unrelated molecules, including many antibiotics, dyes, and steroids. The AcrB inner membrane pump portion of the efflux system has been shown in recent structural studies to bind substrates at multiple sites, suggesting that particular substrate "sets" may compete for efflux by interfering with a certain binding site(s). However, our data indicate that the general structural class does not appear to dictate a particular substrate binding site that can be competitively inhibited in whole cells. In our study, substrate competition failed to increase cell-associated levels of steroids or dyes to levels characteristic of AcrB- or AcrB/EmrAB-deficient genomic mutants or achieved with the pump inhibitor carbonyl cyanide m-chlorophenylhydrazone. In addition, this general observation was sustained even with (i) a cocktail containing seven-pump substrates supplied slightly below their respective wild-type MIC levels, (ii) competing drug substrates of the same structural class (steroids or macrolides), and (iii) hyper-MIC levels of the exogenously supplied agents. Thus, this pump system (and possibly EmrAB-TolC) may have an extraordinary capacity to simultaneously handle multiple-drug substrates that is not necessarily reflected in MIC analyses. In addition, our study has extended the range of substrates recognized by the AcrAB- and EmrAB-TolC systems.
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Reserpine
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet Solution
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Gentian violet, suitable for microscopy
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Josamycin, ≥90% (HPLC)
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)