Merck
  • Inhibition of hepatitis C virus replication by peroxidation of arachidonate and restoration by vitamin E.
Inhibition of hepatitis C virus replication by peroxidation of arachidonate and restoration by vitamin E.
Proceedings of the National Academy of Sciences of the United States of America (2007-11-16)
Hua Huang, Yan Chen, Jin Ye
ABSTRACT
Hepatitis C virus (HCV) is a single-stranded positive-sense RNA virus of the Flaviviridae family. HCV-infected hepatocytes are known to produce reactive oxygen species (ROS), which initiate lipid peroxidation, a reaction that converts polyunsaturated fatty acids, such as arachidonate, into reactive carbonyls that inactivate proteins. To study the effect of lipid peroxidation on HCV replication, we administered arachidonate to Huh7 cells that harbor an HCV replicon (Huh7-K2040 cells). After incubation in medium supplemented with arachidonate but deprived of lipid-soluble antioxidants, the cellular amount of malondialdehyde (MDA), a product of lipid peroxidation, increased markedly in Huh7-K2040 cells but not in parental Huh7 cells that do not harbor an HCV replicon. This increase was followed by a sharp reduction (>95%) in HCV RNA. Both of these events were prevented when cells were treated with vitamin E, a lipid-soluble antioxidant. After prolonged incubation of Huh7-K2040 cells with arachidonate in the absence of lipid-soluble antioxidants, the amount of MDA decreased after the reduction in the amount of HCV RNA. Thus, in the presence of arachidonate and in the absence of lipid-soluble antioxidants, HCV replication induces lipid peroxidation that reduces the amount of HCV RNA. Our results provide a mechanism for the previous observation that polyunsaturated fatty acids inhibit HCV replication [Kapadia SB, Chisari FV (2005) Proc Natl Acad Sci USA 102:2561-2566], and they suggest that these agents may be effective in inhibiting HCV replication in vivo.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Supelco
Linolenic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Oleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g