Merck
  • A tissue-selective nonsteroidal progesterone receptor modulator: 7,9-difluoro-5-(3-methylcyclohex-2-enyl)-2,2,4-trimethyl-1,2-dihydrochromeno[3,4-f]quinoline.
A tissue-selective nonsteroidal progesterone receptor modulator: 7,9-difluoro-5-(3-methylcyclohex-2-enyl)-2,2,4-trimethyl-1,2-dihydrochromeno[3,4-f]quinoline.
Journal of medicinal chemistry (2008-06-17)
Bijan Pedram, Arjan van Oeveren, Dale E Mais, Keith B Marschke, Pieter M Verbost, Marinus B Groen, Lin Zhi
ABSTRACT
The progesterone receptor plays an important role in the female reproductive system. Here we describe the discovery of a new selective progesterone receptor modulator (SPRM). In rats, the lead compound, 7,9-difluoro-5-(3-methylcyclohex-2-enyl)-2,2,4-trimethyl-1,2-dihydrochromeno[3,4- f]quinoline ( 5c), inhibited ovulation and showed full efficacy in uterine and vaginal tissue but was a mixed partial agonist/antagonist in breast tissue. The compound also suppressed ovulation in monkeys, but in contrast to currently approved steroidal PR agonists, it did not suppress estradiol levels.
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesterone, VETRANAL®, analytical standard