Merck
  • Complex roles for reactive astrocytes in the triple transgenic mouse model of Alzheimer disease.
Complex roles for reactive astrocytes in the triple transgenic mouse model of Alzheimer disease.
Neurobiology of aging (2020-03-17)
Océane Guillemaud, Kelly Ceyzériat, Thomas Saint-Georges, Karine Cambon, Fanny Petit, Lucile Ben Haim, Maria-Angeles Carrillo-de Sauvage, Martine Guillermier, Sueva Bernier, Anne-Sophie Hérard, Charlène Joséphine, Alexis Pierre Bémelmans, Emmanuel Brouillet, Philippe Hantraye, Gilles Bonvento, Carole Escartin
ABSTRACT
In Alzheimer disease (AD), astrocytes undergo complex changes and become reactive. The consequences of this reaction are still unclear. To evaluate the net impact of reactive astrocytes in AD, we developed viral vectors targeting astrocytes that either activate or inhibit the Janus kinase-signal transducer and activator of transcription 3 (JAK2-STAT3) pathway, a central cascade controlling astrocyte reaction. We aimed to evaluate whether reactive astrocytes contribute to tau as well as amyloid pathologies in the hippocampus of 3xTg-AD mice, an AD model that develops tau hyper-phosphorylation and amyloid deposition. JAK2-STAT3 pathway-mediated modulation of reactive astrocytes in 25% of the hippocampus of 3xTg-AD mice did not significantly influence tau phosphorylation or amyloid processing and deposition at early, advanced, and terminal disease stage. Interestingly, inhibition of the JAK2-STAT3 pathway in hippocampal astrocytes did not improve spatial memory in the Y maze but it did reduce anxiety in the elevated plus maze. Our unique approach to specifically manipulate reactive astrocytes in situ show they may impact behavioral outcomes without influencing tau or amyloid pathology.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2