Merck
  • Synthesis and biological evaluation of alkoxycoumarins as novel nematicidal constituents.
Synthesis and biological evaluation of alkoxycoumarins as novel nematicidal constituents.
Bioorganic & medicinal chemistry letters (2008-09-17)
Kazuto Takaishi, Minoru Izumi, Naomichi Baba, Kazuyoshi Kawazu, Shuhei Nakajima
ABSTRACT
We synthesized all of the monomethoxycoumarins, 5-alkoxycoumarins and their derivatives, and investigated their nematicidal activity against the phytopathogenic nematode, Bursaphelenchus xylophilus. Among the compounds, 5-ethoxycoumarin showed the highest nematicidal activity. Furthermore, 5-ethoxycoumarin was comparatively harmless against both the brine shrimps, Artemia salina, and the Japanese killifish, Oryzias latipes.
Sigma-Aldrich
7-Methoxycoumarin, ≥98%
Sigma-Aldrich
Coumarin, ≥99% (HPLC)