Merck
  • Carbonic anhydrase activators: activation of the archaeal beta-class (Cab) and gamma-class (Cam) carbonic anhydrases with amino acids and amines.
Carbonic anhydrase activators: activation of the archaeal beta-class (Cab) and gamma-class (Cam) carbonic anhydrases with amino acids and amines.
Bioorganic & medicinal chemistry letters (2008-10-22)
Alessio Innocenti, Sabrina A Zimmerman, Andrea Scozzafava, James G Ferry, Claudiu T Supuran
ABSTRACT
Activation of the archaeal beta-class (Cab) and gamma-class (Cam) carbonic anhydrases (CAs, EC 4.2.1.1) with a series of natural and non-natural amino acids and aromatic/heterocyclic amines has been investigated. Cab, Zn-Cam and Co-Cam showed an activation profile with natural, L- and D-amino acids very different of those of the alpha-class enzymes CA I, II and III. Most of these compounds showed medium efficacy as archaeal CA activators, except for D-Phe and L-Tyr which were effective Cab activators (K(A)s of 10.3-10.5 microM), 2-pyridylmethylamine and 1-(2-aminoethyl)-piperazine which effectively activated Zn-Cam (K(A)s of 10.1-11.4 microM) and serotonin, L-adrenaline and 2-pyridylmethylamine which were the best Co-Cam activators (K(A)s of 0.97-8.9 microM). We prove here that the activation mechanisms of the alpha-, beta-, and gamma-class CAs are similar, although the activation profiles with various compounds differ dramatically between these diverse enzymes.
Sigma-Aldrich
Histamine, ≥97.0%
SAFC
L-Tyrosine
SAFC
L-Histidine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, 99%, FCC