Merck
  • Fibroblast expression of an IκB dominant-negative transgene attenuates renal fibrosis.
Fibroblast expression of an IκB dominant-negative transgene attenuates renal fibrosis.
Journal of the American Society of Nephrology : JASN (2010-09-18)
Tsutomu Inoue, Tsuneo Takenaka, Matsuhiko Hayashi, Toshiaki Monkawa, Jun Yoshino, Kouji Shimoda, Eric G Neilson, Hiromichi Suzuki, Hirokazu Okada
ABSTRACT
It is not clear whether interstitial fibroblasts or tubular epithelial cells are primarily responsible for the profibrotic effects of NF-κB activation during renal fibrogenesis. Here, we crossed mice carrying a conditional IκB dominant-negative transgene (IκBdN) with mice transgenic for cell-specific FSP1.Cre (FSP1(+) fibroblasts) or γGT.Cre (proximal tubular epithelia) and challenged all progeny with unilateral ureteral obstruction. We determined NF-κB activation by nuclear localization of phosphorylated p65 ((p)p65) in renal tissues after 7 days. We observed inhibition of NF-κB activation in interstitial cells and tubular epithelia in obstructed kidneys of FSP1.Cre;IκBdN and γGT.Cre;IκBdN mice, respectively, compared with IκBdN controls (P < 0.05). Deposition of extracellular matrix, however, was significantly lower in the obstructed kidneys of FSP1.Cre;IκBdN mice but not in γGT.Cre;IκBdN mice (P < 0.05). In addition, levels of mRNA encoding the profibrotic PAI-1, fibronectin-EIIIA, and type I (α1) procollagen were significantly lower in obstructed kidneys of FSP1.Cre;IκBdN mice compared with γGT.Cre;IκBdN mice (P < 0.05). Taken together, these data support a profibrotic role for fibroblasts, but not proximal tubular epithelial cells, in modulating NF-κB activation during renal fibrogenesis.
Sigma-Aldrich
Extract-N-Amp Tissue PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications