Merck
  • Neutralizing Antibody and Soluble ACE2 Inhibition of a Replication-Competent VSV-SARS-CoV-2 and a Clinical Isolate of SARS-CoV-2.
Neutralizing Antibody and Soluble ACE2 Inhibition of a Replication-Competent VSV-SARS-CoV-2 and a Clinical Isolate of SARS-CoV-2.
Cell host & microbe (2020-08-01)
James Brett Case, Paul W Rothlauf, Rita E Chen, Zhuoming Liu, Haiyan Zhao, Arthur S Kim, Louis-Marie Bloyet, Qiru Zeng, Stephen Tahan, Lindsay Droit, Ma Xenia G Ilagan, Michael A Tartell, Gaya Amarasinghe, Jeffrey P Henderson, Shane Miersch, Mart Ustav, Sachdev Sidhu, Herbert W Virgin, David Wang, Siyuan Ding, Davide Corti, Elitza S Theel, Daved H Fremont, Michael S Diamond, Sean P J Whelan
ABSTRACT
Antibody-based interventions against SARS-CoV-2 could limit morbidity, mortality, and possibly transmission. An anticipated correlate of such countermeasures is the level of neutralizing antibodies against the SARS-CoV-2 spike protein, which engages with host ACE2 receptor for entry. Using an infectious molecular clone of vesicular stomatitis virus (VSV) expressing eGFP as a marker of infection, we replaced the glycoprotein gene (G) with the spike protein of SARS-CoV-2 (VSV-eGFP-SARS-CoV-2) and developed a high-throughput-imaging-based neutralization assay at biosafety level 2. We also developed a focus-reduction neutralization test with a clinical isolate of SARS-CoV-2 at biosafety level 3. Comparing the neutralizing activities of various antibodies and ACE2-Fc soluble decoy protein in both assays revealed a high degree of concordance. These assays will help define correlates of protection for antibody-based countermeasures and vaccines against SARS-CoV-2. Additionally, replication-competent VSV-eGFP-SARS-CoV-2 provides a tool for testing inhibitors of SARS-CoV-2 mediated entry under reduced biosafety containment.
Sigma-Aldrich
Phosphotungstic acid hydrate, suitable for microscopy
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, ≥90% (HPLC), crystalline