Merck
Bovine erythrocyte superoxide dismutase. Complete amino acid sequence.
The Journal of biological chemistry (1974-11-25)
H M Steinman, V R Naik, J L Abernethy, R L Hill
Sigma-Aldrich
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein