Merck
  • Diallyl Biphenyl-Type Neolignans Have a Pharmacophore of PPARα/γ Dual Modulators.
Diallyl Biphenyl-Type Neolignans Have a Pharmacophore of PPARα/γ Dual Modulators.
Biomolecules & therapeutics (2020-06-25)
Yujia Han, Jingjing Liu, Sungjin Ahn, Seungchan An, Hyejin Ko, Jeayoung C Shin, Sun Hee Jin, Min Won Ki, So Hun Lee, Kang Hyuk Lee, Song Seok Shin, Won Jun Choi, Minsoo Noh
ABSTRACT
Adiponectin secretion-promoting compounds have therapeutic potentials in human metabolic diseases. Diallyl biphenyl-type neolignan compounds, magnolol, honokiol, and 4-O-methylhonokiol, from a Magnolia officinalis extract were screened as adiponectin- secretion promoting compounds in the adipogenic differentiation model of human bone marrow mesenchymal stem cells (hBM-MSCs). In a target identification study, magnolol, honokiol, and 4-O-methylhonokiol were elucidated as PPARα and PPARγ dual modulators. Diallyl biphenyl-type neolignans affected the transcription of lipid metabolism-associated genes in a different way compared to those of specific PPAR ligands. The diallyl biphenyl-type neolignan structure provides a novel pharmacophore of PPARα/γ dual modulators, which may have unique therapeutic potentials in diverse metabolic diseases.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Troglitazone, ≥98% (HPLC)