Merck
  • Isolation and Assessment of a Highly-Active Anti-Inflammatory Exopolysaccharide from Mycelial Fermentation of a Medicinal Fungus Cs-HK1.
Isolation and Assessment of a Highly-Active Anti-Inflammatory Exopolysaccharide from Mycelial Fermentation of a Medicinal Fungus Cs-HK1.
International journal of molecular sciences (2021-03-07)
Long-Qing Li, Ang-Xin Song, Wing-Tak Wong, Jian-Yong Wu
ABSTRACT
The purpose of this work was to fractionate the complex exopolysaccharide (EPS) from a medicinal fungus Ophiocordyceps sinensis Cs-HK1 based on the molecular weight (MW) range and to assess the in vitro anti-inflammatory activity of different EPS fractions in THP-1 cell culture. The lower MW fraction (EPS-LM-1) showed a much higher anti-inflammatory activity. EPS-LM-1 was identified as a heteropolysaccharide consisting of mannose, glucose, and galactose residues with an average MW of 360 kDa. EPS-LM-1 significantly inhibited the lipopolysaccharide-induced inflammatory responses with the effective concentrations for 50% inhibition below 5 µg/mL on a few major proinflammatory markers. With such a notable in vitro anti-inflammatory activity, EPS-LM-1 is a promising candidate for the development of a new anti-inflammation therapy.
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Human IL-1 β ELISA Kit, for serum, plasma, cell culture supernatant and urine