Merck
  • Amino acid based enantiomerically pure 3-substituted benzofused heterocycles: A new class of antithrombotic agents.
Amino acid based enantiomerically pure 3-substituted benzofused heterocycles: A new class of antithrombotic agents.
Bioorganic & medicinal chemistry letters (2009-11-26)
Jitendra Kumar Mishra, Krishnananda Samanta, Manish Jain, Madhu Dikshit, Gautam Panda
ABSTRACT
A diverse group of novel medium ring heterocycles derived from naturally abundant proteinogenic amino acids were evaluated for their potency towards antithrombotic activity. The more potent benzofused oxazepine and oxazocine scaffolds were diversified by incorporating different amino acids at the position number 3. Further the effect of ring size has also been taken into account and it was observed that the eight-membered oxazocines ane more potent compared to the corresponding oxazepines.
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%