Merck
Hypoglycemic Effect of Calea urticifolia (Mill.) DC.
Evidence-based complementary and alternative medicine : eCAM (2021-01-29)
Adolfo Andrade-Cetto, Fernanda Espinoza-Hernández, Gerardo Mata-Torres
ABSTRACT
The onset of type 2 diabetes (T2D) is a consequence of the progressive loss of adequate β-cell insulin secretion, which frequently occurs under a background of insulin resistance. Currently, nearly 13 million Mexicans are living with diabetes. Moreover, due to poor socioeconomic conditions and the cultural idiosyncrasies of the Mexican population, the use of medicinal plants to treat T2D is a common practice in Mexico. In the Mexican state of Hidalgo, we found the traditional use of Calea urticifolia (CU) to treat this disease. To treat T2D, people drink an infusion made from the aerial part of the plant throughout the day. With the aim of investigating whether the infusion at a traditional dose produces a hypoglycemic effect in either the fasting or postprandial state, we measured the effect of the infusion in a hyperglycemic animal model (rats administered streptozotocin (STZ) and nicotinamide (NZ)) by conducting a glucose tolerance test and constructing a blood-glucose curve. We then analyzed whether the observed effect was related to the inhibition of glucose absorption in the gut or the inhibition of hepatic glucose output (HGO) in vivo and in vitro. Furthermore, we confirmed our findings by identifying the potential targets of the infusion via a network pharmacology analysis. Through high-performance liquid chromatography (HPLC) and thin layer chromatography (TLC), we detected a number of compounds in the extract and identified two of them. The plant extract produced a highly significant hypoglycemic effect under fasting conditions and a weak hypoglycemic effect following glucose or sucrose challenge. Although the plant extract blocked only 20% of the alpha-glucosidase enzyme activity in vitro, in the pyruvate tolerance test (which measures the liberation of hepatic glucose), it significantly reduced glucose levels. Furthermore, in vitro, the extract diminished the activity of the glucose-6-phosphatase complex by 90%. In addition, by conducting TLC, we detected the presence of chlorogenic acid and rutin, which have been reported to block HGO. The results presented here provide evidence of the hypoglycemic effect of the traditionally used C. urticifolia extract and demonstrate that this effect is associated with both a reduction in glucose synthesis via gluconeogenesis due to the phytochemical composition of the extract and a slight blockage of glucose absorption in the gut.
Sigma-Aldrich
4-Nitrophenyl α-D-glucopyranoside, ≥99%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Intestinal acetone powders from rat
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%