- Serotonin derivatives as a new class of non-ATP-competitive receptor tyrosine kinase inhibitors.
Serotonin derivatives as a new class of non-ATP-competitive receptor tyrosine kinase inhibitors.
Bioorganic & medicinal chemistry (2010-05-12)
Anita Büttner, Thomas Cottin, Jing Xu, Lito Tzagkaroulaki, Athanassios Giannis
PMID20456957
ABSTRACT
The discovery of new templates and their subsequent elaboration to clinically useful receptor tyrosine kinase (RTK) inhibitors continues to be an important issue. RTKs are a class of enzymes responsible for the activation of different cellular signal transduction cascades. The majority of the known small molecules RTK inhibitors are ATP-competitive and they are multiple targeted inhibitors. We describe here serotonin derivatives as a new class of multiple targeted RTK inhibitors. In contrast to most other RTK inhibitors they act via a non-ATP-competitive (allosteric) mechanism. Furthermore, they are able to inhibit the proliferation of HUVE cells, fibroblasts and two cancer cell lines.