  • Search Results
  • Opioids drive breast cancer metastasis through the δ-opioid receptor and oncogenic STAT3.
Opioids drive breast cancer metastasis through the δ-opioid receptor and oncogenic STAT3.
Neoplasia (New York, N.Y.) (2021-01-20)
Sabrina Tripolt, Heidi A Neubauer, Vanessa M Knab, Dominik P Elmer, Fritz Aberger, Richard Moriggl, Daniela A Fux
ABSTRACT
The opioid crisis of pain medication bears risks from addiction to cancer progression, but little experimental evidence exists. Expression of δ-opioid receptors (DORs) correlates with poor prognosis for breast cancer patients, but mechanistic insights into oncogenic signaling mechanisms of opioid-triggered cancer progression are lacking. We show that orthotopic transplant models using human or murine breast cancer cells displayed enhanced metastasis upon opioid-induced DOR stimulation. Interestingly, opioid-exposed breast cancer cells showed enhanced migration and strong STAT3 activation, which was efficiently blocked by a DOR-antagonist. Furthermore, opioid treatment resulted in down-regulation of E-Cadherin and increased expression of epithelial-mesenchymal transition markers. Notably, STAT3 knockdown or upstream inhibition through the JAK1/2 kinase inhibitor ruxolitinib prevented opioid-induced breast cancer cell metastasis and migration in vitro and in vivo. We conclude on a novel mechanism whereby opioid-triggered breast cancer metastasis occurs via oncogenic JAK1/2-STAT3 signaling to promote epithelial-mesenchymal transition. These findings emphasize the importance of selective and restricted opioid use, as well as the need for safer pain medication that does not activate these oncogenic pathways.
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
[D-Ala2, D-Leu5]-Enkephalin acetate salt, ≥95% (HPLC)
Sigma-Aldrich
11-Amino-1-undecanethiol hydrochloride, 97%