Merck
  • Synthesis of 5-[alkoxy-(4-nitro-phenyl)-methyl]-uridines and study of their cytotoxic activity.
Synthesis of 5-[alkoxy-(4-nitro-phenyl)-methyl]-uridines and study of their cytotoxic activity.
European journal of medicinal chemistry (2010-07-08)
Lucie Brulíková, Petr Dzubák, Marián Hajdúch, Lenka Lachnitová, Madhusudhan Kollareddy, Milan Kolár, Katerina Bogdanová, Jan Hlavác
ABSTRACT
A series of uridine analogues modified at the 5-position with the 5-[alkoxy-(4-nitrophenyl)-methyl] moiety was synthesized. Nucleosides were formed as a mixture of two diastereoisomers, which were separated and tested for their cytotoxic activity in vitro against different cancer cell lines and for antimicrobial activity. Relationships between structure and the above mentioned activities were studied. The cytotoxic activity was slightly increased in some cases by transformation of bases to nucleosides. Depending on the length of the alkyl chain increased cytotoxic and antimicrobial activity were noted. The cytotoxic activity of the nucleosides was not due to cell cycle alterations, DNA and/or RNA synthesis.
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor