Merck
  • Synthesis of new pyrrolo[2,3-d]pyrimidine derivatives as antibacterial and antifungal agents.
Synthesis of new pyrrolo[2,3-d]pyrimidine derivatives as antibacterial and antifungal agents.
European journal of medicinal chemistry (2010-09-11)
Khalid Mohammed Hassan Hilmy, Maha M A Khalifa, Mohammed Abd Allah Hawata, Reda Mohammed Aboalzeen Keshk, Abd Almeneam el-Torgman
ABSTRACT
A series of new pyrrole derivatives, pyrrolo[2,3-d]pyrimidine derivatives, pyrrolotriazolopyrimidines and pyrrolotetrazolopyrimidines were synthesized. The evaluation of their antimicrobial activities against Staphylococcus aureus, Escherichia coli, and Candida albicans were carried out. Pyrrolo[2,3-d]pyrimidines 3a-d, 7a,e, 11d exhibited excellent activity against C. albicans with MIC 0.31-0.62 mg/mL. These compounds displayed better antifungal activity than that of standard drug (fluconazole with MIC 1.5 mg/mL). Furthermore, pyrrolo[2,3-d]pyrimidines 3b,c, 7e exhibited the best activity against S. aureus with MIC 0.31 mg/mL, compared with the standard drug (ampicillin with MIC 0.62 mg/mL). The rest of the compounds were found to be inactive against bacteria and fungi.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)