Merck
  • Nitrogen-Doped Carbons Derived from Imidazole-Based Cross-Linked Porous Organic Polymers.
Nitrogen-Doped Carbons Derived from Imidazole-Based Cross-Linked Porous Organic Polymers.
Molecules (Basel, Switzerland) (2021-01-31)
Wojciech Kiciński, Sławomir Dyjak
ABSTRACT
Nitrogen-doped and heteroatom multi-doped carbon materials are considered excellent metal-free catalysts, superior catalyst supports for transition metal particles and single metal atoms (single-atom catalysts), as well as efficient sorbents for gas- and liquid-phase substances. Acid-catalyzed sol-gel polycondensation of hydroxybenzenes with heterocyclic aldehydes yields cross-linked thermosetting resins in the form of porous organic polymers (i.e., organic gels). Depending on the utilized hydroxybenzene (e.g., phenol, resorcinol, phloroglucinol, etc.) and heterocyclic aldehyde variety of heteroatom-doped organic polymers can be produced. Upon pyrolysis, highly porous and heteroatom-doped carbons are obtained. Herein, polycondensation of phloroglucinol with imidazole-2-carboxaldehyde (and other, similar heterocyclic aldehydes with two heteroatoms in the aromatic ring) is utilized to obtain porous, N-doped organic and carbon gels with N-content of up to 16.5 and 12 wt.%, respectively. Utilization of a heterocyclic aldehyde with two different heteroatoms yields dually-doped carbon materials. Upon pyrolysis, the porous polymers yield ultramicroporous N-doped and N,S co-doped carbons with specific surface areas of up to 800 m2g-1. The influence of the initial composition of reactants and the pyrolysis temperature on the structure and chemical composition of the final doped organic and carbon materials is studied in detail.
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
1-Methyl-2-imidazolecarboxaldehyde, 98%
Sigma-Aldrich
2-Thiazolecarboxaldehyde, 97%
Sigma-Aldrich
2-Imidazolecarboxaldehyde, 97%