Merck
  • Synthesis and anti-tumor activity of novel ethyl 3-aryl-4-oxo-3,3a,4,6-tetrahydro-1H-furo[3,4-c]pyran-3a-carboxylates.
Synthesis and anti-tumor activity of novel ethyl 3-aryl-4-oxo-3,3a,4,6-tetrahydro-1H-furo[3,4-c]pyran-3a-carboxylates.
Bioorganic & medicinal chemistry letters (2011-04-26)
Tiantian Wang, Jia Liu, Hanyu Zhong, Huan Chen, Zhiliang Lv, Yikai Zhang, Mingfeng Zhang, Dongping Geng, Chunjuan Niu, Yongmei Li, Ke Li
ABSTRACT
A series of ethyl 3-aryl-4-oxo-3,3a,4,6-tetrahydro-1H-furo[3,4-c]pyran-3a-carboxylates were prepared through the metal-catalyzed domino reaction of alkylidene malonates and 1,4-butynediol under a one-pot reaction condition at room temperature. Their in vitro anti-proliferative activities were subsequently evaluated in A549, QGY and HeLa cells. The majority of the compounds showed potent anti-tumor activity against HeLa cells. In particular, compound 3l was the most potent compound with IC(50) value of 5.4 μM. For the first time, the X-ray structure of the anti-tumor ethyl 3-aryl-4-oxo-3,3a,4,6-tetrahydro-1H-furo[3,4-c]pyran-3a-carboxylates is determined.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications