Merck
Anti-inflammatory principles from Cordyceps sinensis.
Journal of natural products (2011-08-19)
Mei-Lin Yang, Ping-Chung Kuo, Tsong-Long Hwang, Tian-Shung Wu
ABSTRACT
In order to explore the anti-inflammatory principles of the mycelia of Cordyceps sinensis, the crude extract and partially purified fractions were examined for their inhibition of superoxide anion generation and elastase release. Further chemical investigation of the bioactive fractions has resulted in the identification of 50 compounds, including five constituents, cordysinins A-E (1-5), reported from a natural source for the first time. In addition, compounds were examined for their anti-inflammatory activity. 1-(5-Hydroxymethyl-2-furyl)-β-carboline displayed the most significant inhibition of superoxide anion generation and elastase release with IC50 values of 0.45±0.15 and 1.68±0.32 μM, respectively.
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
2-Furoic acid, 98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Daidzein, ≥98%, synthetic