Merck
  • Co-Adjuvant Therapy Efficacy of Catechin and Procyanidin B2 with Docetaxel on Hormone-Related Cancers In Vitro.
Co-Adjuvant Therapy Efficacy of Catechin and Procyanidin B2 with Docetaxel on Hormone-Related Cancers In Vitro.
International journal of molecular sciences (2021-07-21)
Mª Jesús Núñez-Iglesias, Silvia Novio, Carlota García, Mª Elena Pérez-Muñuzuri, María-Carmen Martínez, José-Luis Santiago, Susana Boso, Pilar Gago, Manuel Freire-Garabal
ABSTRACT
Prostate (PC) and breast cancer (BC) are heterogeneous hormonal cancers. Treatment resistance and adverse effects are the main limitations of conventional chemotherapy treatment. The use of sensitizing agents could improve the effectiveness of chemotherapeutic drugs as well as obviate these limitations. This study analyzes the effect of single catechin (CAT), procyanidin B2 (ProB2) treatment as well as the co-adjuvant treatment of each of these compounds with docetaxel (DOCE). We used PC- and BC-derived cell lines (PC3, DU-145, T47D, MCF-7 and MDA-MB-231). The short and long-term pro-apoptotic, anti-proliferative and anti-migratory effects were analyzed. RT-qPCR was used to discover molecular bases of the therapeutic efficacy of these compounds. ProB2 treatment induced a two- to five-fold increase in anti-proliferative and pro-apoptotic effects compared to single DOCE treatment, and also had a more sensitizing effect than DOCE on DU145 cells. Regarding BC cells, ProB2- and CAT-mediated sensitization to DOCE anti-proliferative and pro-apoptotic effects was cell-independent and cell-dependent, respectively. Combined treatment led to high-efficacy effects on MCF-7 cells, which were associated to the up-regulation of CDKN1A, BAX, caspase 9 and E-cadherin mRNA under combined treatment compared to single DOCE treatment. CAT and ProB2 can enhance the efficacy of DOCE therapy on PC and BC cells by the sensitizing mechanism.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
GenElute mRNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture