Merck
IRGM1 links mitochondrial quality control to autoimmunity.
Nature immunology (2021-01-30)
Prashant Rai, Kyathanahalli S Janardhan, Julie Meacham, Jennifer H Madenspacher, Wan-Chi Lin, Peer W F Karmaus, Jennifer Martinez, Quan-Zhen Li, Mei Yan, Jialiu Zeng, Mark W Grinstaff, Orian S Shirihai, Gregory A Taylor, Michael B Fessler
ABSTRACT
Mitochondrial abnormalities have been noted in lupus, but the causes and consequences remain obscure. Autophagy-related genes ATG5, ATG7 and IRGM have been previously implicated in autoimmune disease. We reasoned that failure to clear defective mitochondria via mitophagy might be a foundational driver in autoimmunity by licensing mitochondrial DNA-dependent induction of type I interferon. Here, we show that mice lacking the GTPase IRGM1 (IRGM homolog) exhibited a type I interferonopathy with autoimmune features. Irgm1 deletion impaired the execution of mitophagy with cell-specific consequences. In fibroblasts, mitochondrial DNA soiling of the cytosol induced cyclic GMP-AMP synthase (cGAS)-stimulator of interferon genes (STING)-dependent type I interferon, whereas in macrophages, lysosomal Toll-like receptor 7 was activated. In vivo, Irgm1-/- tissues exhibited mosaic dependency upon nucleic acid receptors. Whereas salivary and lacrimal gland autoimmune pathology was abolished and lung pathology was attenuated by cGAS and STING deletion, pancreatic pathology remained unchanged. These findings reveal fundamental connections between mitochondrial quality control and tissue-selective autoimmune disease.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
Pepstatin A, microbial, Purity ≥98%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
mTOR Inhibitor XI, Torin1, mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).