Merck
  • Reversible modification of pig heart mitochondrial malate dehydrogenase by pyridoxal 5'-phosphate.
Reversible modification of pig heart mitochondrial malate dehydrogenase by pyridoxal 5'-phosphate.
The Biochemical journal (1975-11-01)
S S Chen, P C Engel
PMID175777
ABSTRACT
1. Pig heart mitochondrial malate dehydrogenase incubated with pyridoxal 5'-phosphate at pH 8.0 and 25 degrees C gradually loses activity. Such inactivation can be largely reversed by dialysis or by addition of L-lysine or L-cysteine, and can be made permanent by NaBH4 reduction. 2. Modification of malate dehydrogenase with pyridoxal 5'-phosphate at 35 degrees C involves two phases, an initial inactivation which is reversible and a slower irreversible second stage. 3. The initial reaction between pyridoxal 5'-phosphate and malate dehydrogenase appears to involve reversible formation of a Schiff base with the epsilon-amino group of a lysine residue. 4. Inactivation of malate dehydrogenase by pyridoxal 5'-phosphate at 10 degrees C involves only the reversible reaction. 5. At 10 degrees C repeated cycles of treatment with pyridoxal 5'-phosphate and NaBH4 reduction lead to a stepwise decline in residual activity. 6. Apparent Km values for malate and NAD+ are unaltered in the partially inactivated enzyme. 7. NAD+ and NADH give only partial protection against pyridoxal 5'-phosphate inactivation. Substrates give no effect.
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Pricing and availability is not currently available.