Merck
  • A second purine nucleoside phosphorylase in Escherichia coli K-12. II. Properties of xanthosine phosphorylase and its induction by xanthosine
A second purine nucleoside phosphorylase in <I>Escherichia coli</I> K-12. II. Properties of xanthosine phosphorylase and its induction by xanthosine
Molecular Genetics and Genomics (1980)
Hmmer-Jespersen, K.