Merck
  • Generation, expansion and functional analysis of endothelial cells and pericytes derived from human pluripotent stem cells.
Generation, expansion and functional analysis of endothelial cells and pericytes derived from human pluripotent stem cells.
Nature protocols (2014-05-31)
Valeria V Orlova, Francijna E van den Hil, Sandra Petrus-Reurer, Yvette Drabsch, Peter Ten Dijke, Christine L Mummery
ABSTRACT
Human endothelial cells (ECs) and pericytes are of great interest for research on vascular development and disease, as well as for future therapy. This protocol describes the efficient generation of ECs and pericytes from human pluripotent stem cells (hPSCs) under defined conditions. Essential steps for hPSC culture, differentiation, isolation and functional characterization of ECs and pericytes are described. Substantial numbers of both cell types can be derived in only 2-3 weeks: this involves differentiation (10 d), isolation (1 d) and 4 or 10 d of expansion of ECs and pericytes, respectively. We also describe two assays for functional evaluation of hPSC-derived ECs: (i) primary vascular plexus formation upon coculture with hPSC-derived pericytes and (ii) incorporation in the vasculature of zebrafish xenografts in vivo. These assays can be used to test the quality and drug sensitivity of hPSC-derived ECs and model vascular diseases with patient-derived hPSCs.
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Monoclonal Anti-Caldesmon (Smooth) antibody produced in mouse, clone hHCD, ascites fluid
Sigma-Aldrich
L-Ascorbic acid 2-phosphate sesquimagnesium salt hydrate, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Diluent C for General Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
PKH67 Green Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Methylene blue, certified by the BSC