Merck
  • IRS-1 is a common element in insulin and insulin-like growth factor-I signaling to the phosphatidylinositol 3'-kinase.
IRS-1 is a common element in insulin and insulin-like growth factor-I signaling to the phosphatidylinositol 3'-kinase.
Endocrinology (1993-04-01)
M G Myers, X J Sun, B Cheatham, B R Jachna, E M Glasheen, J M Backer, M F White
ABSTRACT
IRS-1 is a unique cytosolic protein that becomes tyrosine phosphorylated during insulin stimulation of intact cells and immediately associates with the phosphatidylinositol 3'-kinase (PtdIns 3'-kinase). The insulin-like growth factor-I (IGF-I) receptor also mediated the tyrosine phosphorylation of IRS-1 and increased the amount of PtdIns 3'-kinase activity bound to IRS-1 in Chinese hamster ovary cells. Purified insulin receptor and IGF-I receptor phosphorylated recombinant baculovirus-produced IRS-1 on similar sites in vitro, and phosphorylated baculovirus-produced IRS-1 bound PtdIns 3'-kinase activity from lysates of quiescent cells. Treatment of cells with IGF-I activated the PtdIns 3'-kinase, suggesting that IGF-I activates the PtdIns 3'-kinase through IRS-1 binding to p85 in a manner similar to insulin. Chinese hamster ovary cells overexpressing IRS-1 demonstrated increased tyrosine phosphorylation of IRS-1, and more PtdIns 3'-kinase activity associated with IRS-1 in these cells. These data demonstrate that IRS-1 is a common element for signal transmission by the IGF-I and insulin receptors.