Merck
Quantitative modeling of regular retinal microglia distribution.
Scientific reports (2021-11-24)
Yoshie Endo, Daisuke Asanuma, Shigeyuki Namiki, Kei Sugihara, Kenzo Hirose, Akiyoshi Uemura, Yoshiaki Kubota, Takashi Miura
ABSTRACT
Microglia are resident immune cells in the central nervous system, showing a regular distribution. Advancing microscopy and image processing techniques have contributed to elucidating microglia's morphology, dynamics, and distribution. However, the mechanism underlying the regular distribution of microglia remains to be elucidated. First, we quantitatively confirmed the regularity of the distribution pattern of microglial soma in the retina. Second, we formulated a mathematical model that includes factors that may influence regular distribution. Next, we experimentally quantified the model parameters (cell movement, process formation, and ATP dynamics). The resulting model simulation from the measured parameters showed that direct cell-cell contact is most important in generating regular cell spacing. Finally, we tried to specify the molecular pathway responsible for the repulsion between neighboring microglia.
Sigma-Aldrich
Phospholipase C, Phosphatidylinositol-specific from Bacillus cereus, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate, ≥93% (HPLC), powder