Merck
  • A Quantitative Chemoproteomic Platform to Monitor Selenocysteine Reactivity within a Complex Proteome.
A Quantitative Chemoproteomic Platform to Monitor Selenocysteine Reactivity within a Complex Proteome.
Cell chemical biology (2018-07-10)
Daniel W Bak, Jinjun Gao, Chu Wang, Eranthie Weerapana
ABSTRACT
Mammalian selenocysteine (Sec)-containing proteins, selenoproteins, are important to (patho)physiological processes, including redox homeostasis. Sec residues have been recalcitrant to mass spectrometry-based chemoproteomic methods that enrich for reactive cysteine (Cys) residues with electrophilic chemical probes, despite confirmed reactivity of Sec with these electrophiles. Highly abundant Cys peptides likely suppress low-abundant Sec peptides. By exploiting the decreased pKa of Sec relative to Cys, we have developed a chemoproteomic platform that relies on low pH (pH 5.75) electrophile labeling, reducing Cys reactivity and enhancing identification of Sec-containing peptides across mouse tissues and cell lines. The utility of this Sec-profiling platform is underscored by evaluation of the selectivity of auranofin, an inhibitor of the selenoprotein, thioredoxin reductase, against both reactive Cys- and Sec-containing proteins. Platform limitations pertain to the non-physiological low-pH conditions that could perturb protein structure and function. Future work necessitates the discovery of Sec-selective electrophiles that function at physiological pH.
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%