Merck
  • Oversulfated chondroitin sulfate is a contaminant in heparin associated with adverse clinical events.
Oversulfated chondroitin sulfate is a contaminant in heparin associated with adverse clinical events.
Nature biotechnology (2008-04-26)
Marco Guerrini, Daniela Beccati, Zachary Shriver, Annamaria Naggi, Karthik Viswanathan, Antonella Bisio, Ishan Capila, Jonathan C Lansing, Sara Guglieri, Blair Fraser, Ali Al-Hakim, Nur Sibel Gunay, Zhenqing Zhang, Luke Robinson, Lucinda Buhse, Moheb Nasr, Janet Woodcock, Robert Langer, Ganesh Venkataraman, Robert J Linhardt, Benito Casu, Giangiacomo Torri, Ram Sasisekharan
ABSTRACT
Recently, certain lots of heparin have been associated with an acute, rapid onset of serious side effects indicative of an allergic-type reaction. To identify potential causes for this sudden rise in side effects, we examined lots of heparin that correlated with adverse events using orthogonal high-resolution analytical techniques. Through detailed structural analysis, the contaminant was found to contain a disaccharide repeat unit of glucuronic acid linked beta1-->3 to a beta-N-acetylgalactosamine. The disaccharide unit has an unusual sulfation pattern and is sulfated at the 2-O and 3-O positions of the glucuronic acid as well as at the 4-O and 6-O positions of the galactosamine. Given the nature of this contaminant, traditional screening tests cannot differentiate between affected and unaffected lots. Our analysis suggests effective screening methods that can be used to determine whether or not heparin lots contain the contaminant reported here.