Merck
  • Enzymatic synthesis of nucleosides by nucleoside phosphorylase co-expressed in Escherichia coli
Enzymatic synthesis of nucleosides by nucleoside phosphorylase co-expressed in <I>Escherichia coli</I>
Journal of Zhejiang University. Science (2010)
Ding, Q., et al.