Merck
  • In Vivo Cerebral Translocator Protein (TSPO) Binding and Its Relationship with Blood Adiponectin Levels in Treatment-Naïve Young Adults with Major Depression: A [11C]PK11195 PET Study.
In Vivo Cerebral Translocator Protein (TSPO) Binding and Its Relationship with Blood Adiponectin Levels in Treatment-Naïve Young Adults with Major Depression: A [11C]PK11195 PET Study.
Biomedicines (2022-01-22)
Yo-Han Joo, Min-Woo Lee, Young-Don Son, Keun-A Chang, Maqsood Yaqub, Hang-Keun Kim, Paul Cumming, Jong-Hoon Kim
ABSTRACT
Adiponectin is an adipokine that mediates cellular cholesterol efflux and plays important roles in neuroinflammatory processes. In this study, we undertook positron emission tomography (PET) with the translocator protein (TSPO) ligand [11C]PK11195 and measured serum adiponectin levels in groups of treatment-naïve young adult patients with major depressive disorder (MDD) and matched healthy controls. Thirty treatment-naïve MDD patients (median age: 24 years) and twenty-three healthy controls underwent [11C]PK11195 PET. We quantified TSPO availability in brain as the [11C]PK11195 binding potential (BPND) using a reference tissue model in conjunction with the supervised cluster analysis (SVCA4) algorithm. Age, sex distribution, body mass index, and serum adiponectin levels did not differ between the groups. Between-group analysis using a region-of-interest approach showed significantly higher [11C]PK11195 BPND in the left anterior and right posterior cingulate cortices in MDD patients than in controls. Serum adiponectin levels had significant negative correlations with [11C]PK11195 BPND in the bilateral hippocampus in MDD patients, but significant positive correlations in the bilateral hippocampus in the control group. Our results indicate significantly higher TSPO binding in the anterior and posterior cingulate cortices in treatment-naïve young MDD patients, suggesting microglial activation in these limbic regions, which are involved in cognitive and emotional processing. The opposite correlations between [11C]PK11195 BPND in the hippocampus with serum adiponectin levels in MDD and control groups suggest that microglial activation in the hippocampus may respond differentially to adiponectin signaling in MDD and healthy subjects, possibly with respect to microglial phenotype.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Millipore
Protease Inhibitor Cocktail Set III, Animal-Free, Protease Inhibitor Cocktail Set III, Animal-Free, is a cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine & serine proteases & aminopeptidases.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution