Merck
  • Fabrication of a paper-based colorimetric molecular test for SARS-CoV-2.
Fabrication of a paper-based colorimetric molecular test for SARS-CoV-2.
MethodsX (2021-11-30)
Jiangshan Wang, Andres Dextre, Ana Pascual-Garrigos, Josiah Levi Davidson, Darby McChesney, Jordan Seville, Mohit S Verma
ABSTRACT
The ongoing pandemic caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has caused unprecedented damage to the global economy. Diagnostic testing is a key factor in limiting virus transmission and safeguarding public health. We present the fabrication process of a paper-based device that uses reverse-transcription loop-mediated isothermal amplification (RT-LAMP) to detect SARS-CoV-2 in complex matrices by providing a colorimetric response apparent to the naked eye. Because of LAMP's functionality, this device just requires a simple heat source (e.g., water bath, incubator), it can be deployed in resource-constrained areas and can be used as a supplement to current point-of-care (POC) and community testing procedures. Since the test is based on nucleic acids, the testing platform itself lends to further applications including food safety monitoring, animal diagnostics, etc. simply by changing the specific primers.•We developed a platform capable of on-paper detection of SARS-CoV-2 using colorimetric reporters that produce responses visible to the naked eye.•The platform is easily reconfigurable to target different pathogens by changing the primer sets, and multiplexing is possible by adding additional reaction sites to the device.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent