Merck
  • Oleoylethanolamide restores alcohol-induced inhibition of neuronal proliferation and microglial activity in striatum.
Oleoylethanolamide restores alcohol-induced inhibition of neuronal proliferation and microglial activity in striatum.
Neuropharmacology (2018-11-30)
Patricia Rivera, Daniel Silva-Peña, Eduardo Blanco, Antonio Vargas, Sergio Arrabal, Antonia Serrano, Francisco Javier Pavón, Laura Bindila, Beat Lutz, Fernando Rodríguez de Fonseca, Juan Suárez
ABSTRACT
Previous findings demonstrate a homeostatic role for oleoylethanolamide (OEA) signaling in the ethanol-related neuroinflammation and behavior. However, extensive research is still required in order to unveil the effects of OEA on a number of neurobiological functions such as adult neurogenesis, cell survival and resident neuroimmunity that become notably altered by alcohol. Daily consumption of ethanol (10%) for 2 weeks (6.3 ± 1.1 g/kg/day during last 5 days) caused hypolocomotor activity in rats. This effect appears to rely on central signaling mechanisms given that alcohol increased the OEA levels, the gene expression of OEA-synthesizing enzyme Nape-pld and the number of PPARα-immunoreactive neurons in the striatum. Ethanol-related neurobiological alterations such as a reduction in the number of microglial cells expressing iNOS (a cytokine-inducible immune defense) and in adult neural stem/progenitor cell (NSPC) proliferation (phospho-H3 and BrdU) and maturation (BrdU/β3-tubulin), as well as an increase in damage cell activity (FosB) and apoptosis (cleaved caspase 3) were also observed in the rat striatum. Pharmacological administration of OEA (10 mg/kg) for 5 days during ethanol exposure exacerbated ethanol-induced hypolocomotion and cell apoptosis in the striatum. Interestingly, OEA abrogated the impaired effects of ethanol on PPARα-positive cell population and NSPC proliferation and maturation. OEA also decreased astrocyte-related vimentin immunoreactivity and increased microglial cell population (Iba-1, iNOS) in the striatum. These results suggest that OEA-PPARα signaling modulates glial activation, cell apoptosis and NSPC proliferation and maturation in response to striatal-specific neurobiological alterations induced by prolonged ethanol intake in rats.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
Anti-Iba1/AIF1 Antibody, clone 20A12.1, from mouse
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit