Merck
  • Aryl Hydrocarbon Receptor Activation Produces Heat Shock Protein 90 and 70 Overexpression, Prostaglandin E2/Wnt/β-Catenin Signaling Disruption, and Cell Proliferation in MCF-7 and MDA-MB-231 Cells after 24 h and 14 Days of Chlorpyrifos Treatment.
Aryl Hydrocarbon Receptor Activation Produces Heat Shock Protein 90 and 70 Overexpression, Prostaglandin E2/Wnt/β-Catenin Signaling Disruption, and Cell Proliferation in MCF-7 and MDA-MB-231 Cells after 24 h and 14 Days of Chlorpyrifos Treatment.
Chemical research in toxicology (2021-08-24)
Paula Moyano, José Manuel Garcia, Jimena García, Adela Pelayo, Pilar Muñoz-Calero, María Teresa Frejo, Andrea Flores, Javier Del Pino
ABSTRACT
The biocide chlorpyrifos (CPF) was described to increase breast cancer risk in humans, to produce breast cancer in animals, and to induce cell proliferation in MCF-7 and MDA-MB-231 cells after 1 and 14 days of treatment. The entire mechanisms related to these CPF actions remain unknown. CPF induced cell proliferation in MCF-7 and MDA-MB-231 cells after 1 and 14 days of treatment by AhR activation through the PGE2/Wnt/β-catenin pathway and HSP90 and HSP70 overexpression. Our results reveal new information on CPF toxic mechanisms induced in human breast cancer cell lines, which could assist in elucidating its involvement in breast cancer.