- Designing Successful Proteomics Experiments.
Designing Successful Proteomics Experiments.
Methods in molecular biology (Clifton, N.J.) (2017-02-12)
Daniel Ruderman
PMID28188536
ABSTRACT
Because proteomics experiments are so complex they can readily fail, and do so without clear cause. Using standard experimental design techniques and incorporating quality control can greatly increase the chances of success. This chapter introduces the relevant concepts and provides examples specific to proteomic workflows. Applying these notions to design successful proteomics experiments is straightforward. It can help identify failure causes and greatly increase the likelihood of inter-laboratory reproducibility.
Sigma-Aldrich
Proteomics Dynamic Range Standard Set, Protein Mass Spectrometry Calibration Standard