Merck
  • Methodology to Identify Poly-ADP-Ribose Polymerase 1 (PARP1)-mRNA Targets by PAR-CLiP.
Methodology to Identify Poly-ADP-Ribose Polymerase 1 (PARP1)-mRNA Targets by PAR-CLiP.
Methods in molecular biology (Clifton, N.J.) (2017-07-12)
Manana Melikishvili, Elena Matveeva, Yvonne Fondufe-Mittendorf
ABSTRACT
There is a long list of important RNA-binding proteins (RBP) involved in different steps of gene expression through posttranscriptional modifications: pre-mRNA splicing, mRNA stabilization, polyadenylation, mRNA export from nucleus to the cytoplasm, and translation. The critical role of RNA-protein interaction necessitates a continuous identification of proteins involved in this process. Here we describe the identification of Poly-ADP-Ribose Polymerase 1 (PARP1) as an RNA binding protein involved in RNA splicing.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution