Merck
  • Control of backbone chemistry and chirality boost oligonucleotide splice switching activity.
Control of backbone chemistry and chirality boost oligonucleotide splice switching activity.
Nucleic acids research (2022-01-22)
Pachamuthu Kandasamy, Graham McClorey, Mamoru Shimizu, Nayantara Kothari, Rowshon Alam, Naoki Iwamoto, Jayakanthan Kumarasamy, Gopal R Bommineni, Adam Bezigian, Onanong Chivatakarn, David C D Butler, Michael Byrne, Katarzyna Chwalenia, Kay E Davies, Jigar Desai, Juili Dilip Shelke, Ann F Durbin, Ruth Ellerington, Ben Edwards, Jack Godfrey, Andrew Hoss, Fangjun Liu, Kenneth Longo, Genliang Lu, Subramanian Marappan, Jacopo Oieni, Ik-Hyeon Paik, Erin Purcell Estabrook, Chikdu Shivalila, Maeve Tischbein, Tomomi Kawamoto, Carlo Rinaldi, Joana Rajão-Saraiva, Snehlata Tripathi, Hailin Yang, Yuan Yin, Xiansi Zhao, Cong Zhou, Jason Zhang, Luciano Apponi, Matthew J A Wood, Chandra Vargeese
ABSTRACT
Although recent regulatory approval of splice-switching oligonucleotides (SSOs) for the treatment of neuromuscular disease such as Duchenne muscular dystrophy has been an advance for the splice-switching field, current SSO chemistries have shown limited clinical benefit due to poor pharmacology. To overcome limitations of existing technologies, we engineered chimeric stereopure oligonucleotides with phosphorothioate (PS) and phosphoryl guanidine-containing (PN) backbones. We demonstrate that these chimeric stereopure oligonucleotides have markedly improved pharmacology and efficacy compared with PS-modified oligonucleotides, preventing premature death and improving median survival from 49 days to at least 280 days in a dystrophic mouse model with an aggressive phenotype. These data demonstrate that chemical optimization alone can profoundly impact oligonucleotide pharmacology and highlight the potential for continued innovation around the oligonucleotide backbone. More specifically, we conclude that chimeric stereopure oligonucleotides are a promising splice-switching modality with potential for the treatment of neuromuscular and other genetic diseases impacting difficult to reach tissues such as the skeletal muscle and heart.
Product Description
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Interferon-γ Protein, Recombinant mouse, The Interferon-gamma protein (or IFN-gamma protein) is a regulatory protein produced by activated NK cells & CD4+TCRalpha/beta+, CD8+TCRalpha/beta+ & TCRgamma/delta+ T cells.